Gene Transcripts

Gene Symbol
Fv1
Entrez Gene ID
308568
Gene Name
Friend virus susceptibility 1
Gene Aliases
RGD1309326
Chromosome Location
Chr. 1: 100220789 - 100226728 on Build Rnor_6.0
UniGene ID
Rn.145001
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001014018.2 NP_001014040.2 6 498
XM_008759352.1 XP_008757574.1 5 1878
XM_008759353.1 XP_008757575.1 6 1944
XM_008759354.1 XP_008757576.1 6 1088
XM_008759355.1 XP_008757577.1 7 1154
XM_008759356.1 XP_008757578.1 6 1511
XM_008759357.1 XP_008757579.1 6 1508
XM_008759358.1 XP_008757580.1 6 1485
XM_008759359.1 XP_008757581.1 6 1476
XM_008759360.1 XP_008757582.1 6 716
XM_008759361.1 XP_008757583.1 7 686
XM_008759362.1 XP_008757584.1 7 614
XM_008759363.1 XP_008757585.1 7 782
XM_008759364.1 XP_008757586.1 6 537
XM_008759365.1 XP_008757587.1 7 605
GenBank mRNA BC060592.1 - 6 502
FQ220903.1 - 1 502

View Full Product Details