Gene Transcripts

Gene Symbol
PIP4K2B
Entrez Gene ID
8396
Gene Name
phosphatidylinositol-5-phosphate 4-kinase type 2 beta
Gene Aliases
PI5P4KB, PIP5K2B, PIP5KIIB, PIP5KIIbeta, PIP5P4KB
Chromosome Location
Chr. 17: 38765691 - 38799965 on Build GRCh38
UniGene ID
Hs.260603
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_003559.4 NP_003550.1 3 756
XM_011525326.2 XP_011523628.1 3 816
XM_011525327.2 XP_011523629.1 4 335
XM_011525329.2 XP_011523631.1 3 816
XM_017025196.1 XP_016880685.1 4 333
XM_017025197.1 XP_016880686.1 2 66
XM_017025199.1 XP_016880688.1 3 166
GenBank mRNA BC027459.1 - 3 373
BT019614.1 - 3 275
U85245.1 - 3 755

View Full Product Details