Gene Transcripts

Gene Symbol
Znhit2
Entrez Gene ID
309177
Gene Name
zinc finger, HIT-type containing 2
Gene Aliases
RGD1307918
Chromosome Location
Chr. 1: 221424383 - 221425664 on Build Rnor_6.0
UniGene ID
Rn.18470
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107574.1 NP_001101044.1 1 178
GenBank mRNA BC168685.1 - 1 151

View Full Product Details