Gene Transcripts

Gene Symbol
Olr1687
Entrez Gene ID
309574
Gene Name
olfactory receptor 1687
Gene Aliases
Chromosome Location
Chr. 20: 649582 - 650547 on Build Rnor_6.0
UniGene ID
Rn.107054
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001000508.1 NP_001000508.1 1 641
GenBank mRNA AF091563.1 - 1 440

View Full Product Details