Gene Transcripts

Gene Symbol
Mtmr12
Entrez Gene ID
310155
Gene Name
myotubularin related protein 12
Gene Aliases
Pip3ap
Chromosome Location
Chr. 2: 62236499 - 62305995 on Build Rnor_6.0
UniGene ID
Rn.214625
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001012077.1 NP_001012077.1 5 499
XM_006232046.2 XP_006232108.1 5 590
GenBank mRNA BC089876.1 - 5 499

View Full Product Details