Gene Transcripts
- Gene Symbol
- Qrfpr
- Entrez Gene ID
- 310327
- Gene Name
- pyroglutamylated RFamide peptide receptor
- Gene Aliases
- AQ27, Gpr103
- Chromosome Location
- Chr. 2: 122891321 - 122949241 on Build Rnor_6.0
- UniGene ID
- Rn.162584
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_198199.1 |
NP_937842.1
|
5, 6 |
893 |
|
GenBank mRNA |
AB109630.1 |
- |
5, 6 |
893 |
|
View Full Product Details