Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001257197.1 | NP_001244126.1 | 2 | 542 | |
NM_001257198.1 | NP_001244127.1 | 3 | 423 | ||
NM_003590.4 | NP_003581.1 | 3 | 740 | ||
XM_006712800.2 | XP_006712863.2 | 3 | 413 | ||
XM_011511995.1 | XP_011510297.1 | 3 | 325 | ||
XM_011511996.2 | XP_011510298.1 | 3 | 389 | ||
GenBank mRNA | AB014517.2 | - | 3 | 668 | |
AF062537.1 | - | 3 | 441 | ||
AF064087.1 | - | 3 | 433 | ||
AK291151.1 | - | 3 | 390 | ||
AK299644.1 | - | 2 | 150 | ||
AK308496.1 | - | 3 | 423 | ||
AY337761.1 | - | 3 | 886 | ||
BC039598.1 | - | 3 | 685 | 5784147 | |
BC092409.1 | - | 3 | 685 | 6168545 |