Gene Transcripts

Gene Symbol
Stam2
Entrez Gene ID
311030
Gene Name
signal transducing adaptor molecule (SH3 domain and ITAM motif) 2
Gene Aliases
Chromosome Location
Chr. 3: 38230676 - 38277433 on Build Rnor_6.0
UniGene ID
Rn.161793
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001012085.1 NP_001012085.1 9 930
GenBank mRNA BC083912.1 - 9 930

View Full Product Details