Gene Transcripts
- Gene Symbol
- Zfp334
- Entrez Gene ID
- 311639
- Gene Name
- zinc finger protein 334
- Gene Aliases
- OC-STAMP
- Chromosome Location
- Chr. 3: 162073573 - 162080486 on Build Rnor_6.0
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
XM_230857.7 |
XP_230857.6
|
2 |
2093 |
|
View Full Product Details