Gene Transcripts

Gene Symbol
Il36rn
Entrez Gene ID
311783
Gene Name
interleukin 36 receptor antagonist
Gene Aliases
Il1f5
Chromosome Location
Chr. 3: 1385256 - 1392275 on Build Rnor_6.0
UniGene ID
Rn.63170
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107814.1 NP_001101284.1 6 660
XM_008761586.1 XP_008759808.1 4 1311
XM_008761587.1 XP_008759809.1 5 954

View Full Product Details