Gene Transcripts

Gene Symbol
Ptges2
Entrez Gene ID
311865
Gene Name
prostaglandin E synthase 2
Gene Aliases
Chromosome Location
Chr. 3: 11424195 - 11431379 on Build Rnor_6.0
UniGene ID
Rn.39290
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107832.1 NP_001101302.1 4 1124
GenBank mRNA FQ231959.1 - 4 617

View Full Product Details