Gene Transcripts

Gene Symbol
Slc7a13
Entrez Gene ID
313089
Gene Name
solute carrier family 7 (anionic amino acid transporter), member 13
Gene Aliases
Chromosome Location
Chr. 5: 33784757 - 33805446 on Build Rnor_6.0
UniGene ID
Rn.22708
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001012100.1 NP_001012100.1 1 696
GenBank mRNA BC085796.1 - 1 696

View Full Product Details