Gene Transcripts

Gene Symbol
Cfap221
Entrez Gene ID
313149
Gene Name
cilia and flagella associated protein 221
Gene Aliases
Pcdp1
Chromosome Location
Chr. 13: 35952954 - 36024371 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008769506.1 XP_008767728.1 20 2422

View Full Product Details