Gene Transcripts

Gene Symbol
PPFIA3
Entrez Gene ID
8541
Gene Name
PTPRF interacting protein alpha 3
Gene Aliases
LPNA3
Chromosome Location
Chr. 19: 49119389 - 49151030 on Build GRCh38
UniGene ID
Hs.413748
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_003660.3 NP_003651.1 18 2640
NR_103842.1 17 2417
XM_017027407.1 XP_016882896.1 18 2640
GenBank mRNA AB014554.2 - 18 2623
AF034800.1 - 1 38
AK025972.1 - 15 1840
AK289757.1 - 18 2345
AK293798.1 - 15 1868
BC101518.1 - 18 2366 8069024
BC101520.1 - 18 2366 8069026
BC143470.1 - 17 2143

View Full Product Details