Gene Transcripts

Gene Symbol
Rnf19b
Entrez Gene ID
313806
Gene Name
ring finger protein 19B
Gene Aliases
Ibrdc3
Chromosome Location
Chr. 5: 147246032 - 147270892 on Build Rnor_6.0
UniGene ID
Rn.229171
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001108003.1 NP_001101473.1 6, 7 968
XM_008764147.1 XP_008762369.1 8, 9 1535

View Full Product Details