Gene Transcripts

Gene Symbol
Amn
Entrez Gene ID
314459
Gene Name
amnion associated transmembrane protein
Gene Aliases
Chromosome Location
Chr. 6: 135724455 - 135731896 on Build Rnor_6.0
UniGene ID
Rn.7787
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001108061.1 NP_001101531.1 11 1295
XM_006240588.2 XP_006240650.1 11 1278

View Full Product Details