Gene Transcripts

Gene Symbol
Lrp12
Entrez Gene ID
314941
Gene Name
low density lipoprotein receptor-related protein 12
Gene Aliases
Chromosome Location
Chr. 7: 78859784 - 78930832 on Build Rnor_6.0
UniGene ID
Rn.17834
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001134883.1 NP_001128355.1 5 976
XM_006241606.2 XP_006241668.1 4 872

View Full Product Details