Gene Transcripts

Gene Symbol
RUVBL1
Entrez Gene ID
8607
Gene Name
RuvB like AAA ATPase 1
Gene Aliases
ECP-54, ECP54, INO80H, NMP 238, NMP238, PONTIN, Pontin52, RVB1, TIH1, TIP49, TIP49A
Chromosome Location
Chr. 3: 128064612 - 128153914 on Build GRCh38
UniGene ID
Hs.272822
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001319084.1 NP_001306013.1 6 780
NM_001319086.1 NP_001306015.1 6 1188
NM_003707.2 NP_003698.1 6 721
XM_005247841.3 XP_005247898.1 6 642
XM_011513248.1 XP_011511550.1 6 768
XM_011513249.2 XP_011511551.1 6 775
XM_017007356.1 XP_016862845.1 6 775
XM_017007357.1 XP_016862846.1 6 645
GenBank mRNA AB012122.1 - 6 670
AB451224.1 - 6 617
AB451347.1 - 6 617
AF070735.1 - 6 693
AF099084.1 - 6 681
AJ010058.1 - 6 693
AK222563.1 - 6 702
AK222575.1 - 6 693
AK312290.1 - 6 717
BC002993.1 - 6 688 2823568
BC012886.1 - 6 650 3876400
BT007057.1 - 6 617
DQ469310.1 - 6 617
Y18418.1 - 6 713

View Full Product Details