Gene Transcripts

Gene Symbol
CDK13
Entrez Gene ID
8621
Gene Name
cyclin dependent kinase 13
Gene Aliases
CDC2L, CDC2L5, CHED, hCDK13
Chromosome Location
Chr. 7: 39949501 - 40097134 on Build GRCh38
UniGene ID
Hs.233552
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_003718.4 NP_003709.3 9 3044
NM_031267.3 NP_112557.2 9 3044
XM_017012750.1 XP_016868239.1 10 3993
XM_017012751.1 XP_016868240.1 10 3993
GenBank mRNA AB058694.1 - 3 2460
AJ297709.1 - 9 2894
AJ297710.1 - 9 2894
AK223053.1 - 3 197
BC001274.1 - 8 1314
BF707421.1 - 2 91
M80629.1 - 9 1691

View Full Product Details