Gene Transcripts

Gene Symbol
Nlrx1
Entrez Gene ID
315599
Gene Name
NLR family member X1
Gene Aliases
RGD1311293
Chromosome Location
Chr. 8: 48583559 - 48600203 on Build Rnor_6.0
UniGene ID
Rn.12444
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001025010.1 NP_001020181.1 8 2181
GenBank mRNA BC089834.1 - 8 2181

View Full Product Details