Gene Transcripts

Gene Symbol
Fem1a
Entrez Gene ID
316131
Gene Name
fem-1 homolog a (C. elegans)
Gene Aliases
Chromosome Location
Chr. 9: 10798934 - 10801140 on Build Rnor_6.0
UniGene ID
Rn.228392
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001025706.1 NP_001020877.1 1 958
GenBank mRNA BC097490.1 - 1 958

View Full Product Details