Gene Transcripts

Gene Symbol
Diaph2
Entrez Gene ID
317189
Gene Name
diaphanous-related formin 2
Gene Aliases
Diap2, RGD1564399
Chromosome Location
Chr. X: 99727979 - 100021389 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008773449.1 XP_008771671.1 12 1193
XM_008773450.1 XP_008771672.1 12 1198
XM_008773451.1 XP_008771673.1 11 1172

View Full Product Details