Gene Transcripts

Gene Symbol
Taar8b
Entrez Gene ID
319106
Gene Name
trace amine-associated receptor 8b
Gene Aliases
Ta7
Chromosome Location
Chr. 1: 23565551 - 23568822 on Build Rnor_6.0
UniGene ID
Rn.162405
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175601.1 NP_783191.1 1 905
XM_006227718.2 XP_006227780.2 2 971
XM_006227719.2 XP_006227781.1 2 1393
XM_008758615.1 XP_008756837.1 3 1551

View Full Product Details