Gene Transcripts

Gene Symbol
Zc3h7a
Entrez Gene ID
360466
Gene Name
zinc finger CCCH type containing 7 A
Gene Aliases
Zc3hdc7
Chromosome Location
Chr. 10: 4535650 - 4575643 on Build Rnor_6.0
UniGene ID
Rn.15835
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001108262.1 NP_001101732.1 20 2340
XM_008767489.1 XP_008765711.1 20 3087
XM_008767490.1 XP_008765712.1 21 3281

View Full Product Details