Gene Transcripts

Gene Symbol
Alg1
Entrez Gene ID
360475
Gene Name
ALG1, chitobiosyldiphosphodolichol beta-mannosyltransferase
Gene Aliases
Chromosome Location
Chr. 10: 10539930 - 10550174 on Build Rnor_6.0
UniGene ID
Rn.8024
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001108264.1 NP_001101734.1 3 345
XM_006245802.2 XP_006245864.1 3 381
XM_008767491.1 XP_008765713.1 3 187
XM_008767492.1 XP_008765714.1 2 1245

View Full Product Details