Gene Transcripts

Gene Symbol
B4GALT3
Entrez Gene ID
8703
Gene Name
beta-1,4-galactosyltransferase 3
Gene Aliases
beta4Gal-T3
Chromosome Location
Chr. 1: 161171310 - 161177968 on Build GRCh38
UniGene ID
Hs.321231
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001199873.1 NP_001186802.1 5 1388
NM_001199874.1 NP_001186803.1 5 1317
NM_003779.3 NP_003770.1 5 1367
XM_005245566.1 XP_005245623.1 6 1089
XM_011510093.1 XP_011508395.1 5 923
XM_017002714.1 XP_016858203.1 5 944
XM_017002715.1 XP_016858204.1 5 923
GenBank mRNA AB024435.1 - 3 666
AK023311.1 - 6 852
AK056851.1 - 4 1924
AK092493.1 - 6 849
AK291455.1 - 5 921
BC000276.1 - 5 870 3357342
BC006099.1 - 5 884 3677118
BC009985.2 - 5 846 4124756
Y12509.1 - 3 661

View Full Product Details