Gene Transcripts

Gene Symbol
Cdr2l
Entrez Gene ID
360656
Gene Name
cerebellar degeneration-related protein 2-like
Gene Aliases
RGD1306881
Chromosome Location
Chr. 10: 103934357 - 103948318 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006247771.2 XP_006247833.1 3, 4 798
XM_006247772.1 XP_006247834.1 3, 4 697
XM_008768410.1 XP_008766632.1 3, 4 517
XM_340933.7 XP_340934.5 3, 4 799

View Full Product Details