Gene Transcripts

Gene Symbol
Zfp358
Entrez Gene ID
360754
Gene Name
zinc finger protein 358
Gene Aliases
Chromosome Location
Chr. 12: 2046472 - 2050567 on Build Rnor_6.0
UniGene ID
Rn.22648
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001108328.1 NP_001101798.1 2 370
XM_006248767.1 XP_006248829.1 2 277

View Full Product Details