Gene Transcripts

Gene Symbol
RAB11A
Entrez Gene ID
8766
Gene Name
RAB11A, member RAS oncogene family
Gene Aliases
YL8
Chromosome Location
Chr. 15: 65869459 - 65891991 on Build GRCh38
UniGene ID
Hs.321541
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001206836.1 NP_001193765.1 5 616
NM_004663.4 NP_004654.1 5 711
GenBank mRNA AF000231.1 - 5 651
AF498946.1 - 5 584
AK297549.1 - 5 722
AK298961.1 - 5 504
AK300008.1 - 5 534
AK311770.1 - 5 695
BC013348.2 - 5 678 3510339
BT020151.1 - 5 584
BT020154.1 - 5 584
CR407669.1 - 5 584
CR536493.1 - 5 584
X53143.1 - 5 584
X56740.1 - 5 637

View Full Product Details