Gene Transcripts

Gene Symbol
Cnih2
Entrez Gene ID
361705
Gene Name
cornichon family AMPA receptor auxiliary protein 2
Gene Aliases
CNIH-2, Cnih3, RGD1304930
Chromosome Location
Chr. 1: 220474485 - 220480740 on Build Rnor_6.0
UniGene ID
Rn.18785
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001025132.1 NP_001020303.1 1, 2 336
XM_008760154.1 XP_008758376.1 1, 2 944
GenBank mRNA BC091325.1 - 1, 2 336

View Full Product Details