Gene Transcripts

Gene Symbol
Rfesd
Entrez Gene ID
361871
Gene Name
Rieske (Fe-S) domain containing
Gene Aliases
RGD1308284
Chromosome Location
Chr. 2: 2758382 - 2784368 on Build Rnor_6.0
UniGene ID
Rn.38227
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001108540.1 NP_001102010.1 1, 2 171
XM_006231706.2 XP_006231768.1 2, 3 1271
XM_006231708.2 XP_006231770.1 2, 3 700
XM_006231709.2 XP_006231771.1 2, 3 385
XM_006231711.2 XP_006231773.1 2, 3 1277

View Full Product Details