Gene Transcripts

Gene Symbol
Morn5
Entrez Gene ID
362122
Gene Name
MORN repeat containing 5
Gene Aliases
LRRGT00041, RGD1307739
Chromosome Location
Chr. 3: 15379109 - 15407252 on Build Rnor_6.0
UniGene ID
Rn.57435
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001047912.1 NP_001041377.1 4 368
GenBank mRNA AY383696.1 - 4 368

View Full Product Details