Gene Transcripts

Gene Symbol
Chac1
Entrez Gene ID
362196
Gene Name
ChaC glutathione-specific gamma-glutamylcyclotransferase 1
Gene Aliases
RGD1307153
Chromosome Location
Chr. 3: 111160205 - 111163425 on Build Rnor_6.0
UniGene ID
Rn.23367
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001173437.1 NP_001166908.1 3 720
GenBank mRNA EF688602.1 - 3 720

View Full Product Details