Gene Transcripts

Gene Symbol
Dnajc22
Entrez Gene ID
362998
Gene Name
DnaJ (Hsp40) homolog, subfamily C, member 22
Gene Aliases
RGD1311098
Chromosome Location
Chr. 7: 140781692 - 140786929 on Build Rnor_6.0
UniGene ID
Rn.19564
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001014204.1 NP_001014226.1 2 358
XM_006257363.2 XP_006257425.1 2 620
GenBank mRNA BC086949.1 - 2 358

View Full Product Details