Gene Transcripts

Gene Symbol
Ccdc115
Entrez Gene ID
363213
Gene Name
coiled-coil domain containing 115
Gene Aliases
RGD1304653
Chromosome Location
Chr. 9: 37774876 - 37778839 on Build Rnor_6.0
UniGene ID
Rn.17453
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001108793.1 NP_001102263.1 3 701
GenBank mRNA BC166583.1 - 266

View Full Product Details