Gene Transcripts

Gene Symbol
Rps24
Entrez Gene ID
81776
Gene Name
ribosomal protein S24
Gene Aliases
Chromosome Location
Chr. 16: 757391 - 762091 on Build Rnor_6.0
UniGene ID
Rn.979
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006252533.2 XP_006252595.1 4, 5 493
GenBank mRNA FQ222799.1 - 2, 3 196
FQ222961.1 - 2, 3 196
FQ228664.1 - 2, 3 196

View Full Product Details