Gene Transcripts

Gene Symbol
Ugt1a9
Entrez Gene ID
396552
Gene Name
UDP glucuronosyltransferase 1 family, polypeptide A9
Gene Aliases
Ugt1a10, Ugt1a11
Chromosome Location
Chr. 9: 95161157 - 95302822 on Build Rnor_6.0
UniGene ID
Rn.26489
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_201425.2 NP_958828.1 1 337
GenBank mRNA AY435129.1 - 1 337

View Full Product Details