Gene Transcripts

Gene Symbol
Olr445
Entrez Gene ID
405936
Gene Name
olfactory receptor 445
Gene Aliases
Chromosome Location
Chr. 3: 72997805 - 72998749 on Build Rnor_6.0
UniGene ID
Rn.142985
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001001049.1 NP_001001049.1 1 322

View Full Product Details