Gene Transcripts

Gene Symbol
Prm3
Entrez Gene ID
442921
Gene Name
protamine 3
Gene Aliases
Chromosome Location
Chr. 10: 4951557 - 4952071 on Build Rnor_6.0
UniGene ID
Rn.126718
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001002855.2 NP_001002855.1 1 200
GenBank mRNA S79939.1 - 1 200

View Full Product Details