Gene Transcripts

Gene Symbol
C5ar2
Entrez Gene ID
445269
Gene Name
complement component 5a receptor 2
Gene Aliases
C5L2, Gpr77
Chromosome Location
Chr. 1: 78165155 - 78183413 on Build Rnor_6.0
UniGene ID
Rn.127117
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001003710.2 NP_001003710.2 2 586
XM_006228352.2 XP_006228414.1 2 968
XM_006228353.2 XP_006228415.1 2 740
GenBank mRNA AY600435.1 - 1 452
AY741660.1 - 2 586

View Full Product Details