Gene Transcripts

Gene Symbol
Zfp2
Entrez Gene ID
497897
Gene Name
zinc finger protein 2
Gene Aliases
Mkr-2, Mkr2
Chromosome Location
Chr. 10: 36387135 - 36406789 on Build Rnor_6.0
UniGene ID
Rn.216113
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001127558.1 NP_001121030.1 5 1824
XM_008767695.1 XP_008765917.1 6 1888
XM_008767697.1 XP_008765919.1 6 1896
XM_008767698.1 XP_008765920.1 6 1849
XM_008767699.1 XP_008765921.1 6 1852
GenBank mRNA BC161925.1 - 5 1824

View Full Product Details