Gene Transcripts

Gene Symbol
LOC498339
Entrez Gene ID
498339
Gene Name
LRRGT00190
Gene Aliases
Chromosome Location
Chr. 14: 20456099 - 20474728 on Build Rnor_6.0
UniGene ID
Rn.123784
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001047928.1 NP_001041393.1 4 528
GenBank mRNA AY539941.1 - 4 528

View Full Product Details