Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001017504.1 | NP_001017504.1 | 1 | 275 | |
XM_008770435.1 | XP_008768657.1 | 2 | 2194 | ||
XM_008770436.1 | XP_008768658.1 | 2 | 2118 | ||
GenBank mRNA | BC086322.1 | - | 1 | 275 |