Gene Transcripts

Gene Symbol
Pcdhb2
Entrez Gene ID
498843
Gene Name
protocadherin beta 2
Gene Aliases
RGD1563132
Chromosome Location
Chr. 18: 30375798 - 30378194 on Build Rnor_6.0
UniGene ID
Rn.140789
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001109123.1 NP_001102593.1 1 1382

View Full Product Details