Gene Transcripts

Gene Symbol
Gpr150
Entrez Gene ID
499486
Gene Name
G protein-coupled receptor 150
Gene Aliases
RGD1564729
Chromosome Location
Chr. 2: 2812226 - 2815023 on Build Rnor_6.0
UniGene ID
Rn.133527
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001109173.1 NP_001102643.1 1 232
XM_006231713.2 XP_006231775.1 1 565

View Full Product Details