Gene Transcripts

Gene Symbol
TRIP10
Entrez Gene ID
9322
Gene Name
thyroid hormone receptor interactor 10
Gene Aliases
CIP4, HSTP, STOT, STP, TRIP-10
Chromosome Location
Chr. 19: 6739682 - 6751526 on Build GRCh38
UniGene ID
Hs.515094
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001288962.1 NP_001275891.1 5, 6 490
NM_001288963.1 NP_001275892.1 5, 6 490
NM_004240.3 NP_004231.1 5, 6 490
NR_110231.1 5, 6 490
XM_005259683.2 XP_005259740.1 5, 6 490
XM_006722940.1 XP_006723003.1 5, 6 490
GenBank mRNA AB007845.1 - 5, 6 465
AB072596.1 - 5, 6 465
AF380114.1 - 5, 6 410
AF502289.1 - 5, 6 449
AJ000414.1 - 5, 6 449
AK223109.1 - 5, 6 455
AK313296.1 - 5, 6 472
AL832186.1 - 5, 6 576
AY081141.1 - 5, 6 444
BC013002.2 - 5, 6 435 3532036
BT006698.1 - 5, 6 410
BT020167.1 - 5, 6 410
BT020171.1 - 5, 6 410
CB997985.1 - 5, 6 503
CR536513.1 - 5, 6 410
DB198268.1 - 5, 6 492

View Full Product Details