Gene Transcripts

Gene Symbol
Pglyrp3b
Entrez Gene ID
295180
Gene Name
peptidoglycan recognition protein 3b
Gene Aliases
Chromosome Location
Chr. 2: 191900186 - 191915728 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006232841.2 XP_006232903.1 6 982
XM_006232842.2 XP_006232904.1 6 982
XM_006232844.2 XP_006232906.1 5 829
XM_008761252.1 XP_008759474.1 6 982

Gene Symbol
Pglyrp3
Entrez Gene ID
499658
Gene Name
peptidoglycan recognition protein 3
Gene Aliases
Chromosome Location
Chr. 2: 192253687 - 192264848 on Build Rnor_6.0
UniGene ID
Rn.109236
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001191961.1 NP_001178890.1 852
XM_006232834.2 XP_006232896.1 6 982
XM_006232836.2 XP_006232898.1 5 829

View Full Product Details