Gene Transcripts

Gene Symbol
TGM5
Entrez Gene ID
9333
Gene Name
transglutaminase 5
Gene Aliases
PSS2, TGASE5, TGASEX, TGM6, TGMX, TGX
Chromosome Location
Chr. 15: 43232595 - 43266857 on Build GRCh38
UniGene ID
Hs.129719
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_004245.3 NP_004236.1 12 1855
NM_201631.3 NP_963925.2 13 2101
XM_011522230.2 XP_011520532.1 7 1283
XM_017022729.1 XP_016878218.1 7 1307
GenBank mRNA AF035960.1 - 13 2101
AF035961.1 - 12 1855
AK298479.1 - 9 1517
BC119009.1 - 13 2101 40085374
BC122859.1 - 13 2101 40085372

View Full Product Details