Gene Transcripts

Gene Symbol
Rab43
Entrez Gene ID
500249
Gene Name
RAB43, member RAS oncogene family
Gene Aliases
Chromosome Location
Chr. 4: 119695012 - 119714302 on Build Rnor_6.0
UniGene ID
Rn.141443
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001024331.1 NP_001019502.1 2 361
GenBank mRNA AY593256.1 - 2 361

View Full Product Details