Gene Transcripts

Gene Symbol
Ier3ip1
Entrez Gene ID
500270
Gene Name
immediate early response 3 interacting protein 1
Gene Aliases
Chromosome Location
Chr. 4: 132140802 - 132151787 on Build Rnor_6.0
UniGene ID
Rn.118089
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001047956.1 NP_001041421.1 4 334
XM_008763163.1 XP_008761385.1 3 329
GenBank mRNA AY325263.1 - 4 334

View Full Product Details